missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AKAP5 Full-length ORF (NP_004848.2, 1 a.a. - 427 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009495-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16133742

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein binds to the RII-beta regulatory subunit of PKA, and also to protein kinase C and the phosphatase calcineurin. It is predominantly expressed in cerebral cortex and may anchor the PKA protein at postsynaptic densities (PSD) and be involved in the regulation of postsynaptic events. It is also expressed in T lymphocytes and may function to inhibit interleukin-2 transcription by disrupting calcineurin-dependent dephosphorylation of NFAT. [provided by RefSeq]

Sequence: METTISEIHVENKDEKRSAEGSPGAERQKEKASMLCFKRRKKAAKALKPKAGSEAADVARKCPQEAGASDQPEPTRGAWASLKRLVTRRKRSESSKQQKPLEGEMQPAINAEDADLSKKKAKSRLKIPCIKFPRGPKRSNHSKIIEDSDCSIKVQEEAEILDIQTQTPLNDQATKAKSTQDLSEGISRKDGDEVCESNVSNSITSGEKVISVELGLDNGHSAIQTGTLILEEIETIKEKQDVQPQQASPLETSETDHQQPVLSDVPPLPAIPDQQIVEEASNSTLESAPNGKDYESTEIVAEETKPKDTELSQESDFKENGITEEKSKSEESKRMEPIAIIITDTEISEFDVTKSKNVPKQFLISAENEQVGVFANDNGFEDRTSEQYETLLIETASSLVKNAIQLSIEQLVNEMASDDNKINNLLQ
Specifications

Specifications

NP_004848.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
AKAP5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
9495
AKAP5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AKAP75/AKAP79/H21
AKAP5
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers