Learn More
Abnova™ Human AKAP13 Partial ORF (NP_006729, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011214-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternative splicing of this gene results in at least 3 transcript variants encoding different isoforms containing a dbl oncogene homology (DH) domain and a pleckstrin homology (PH) domain. The DH domain is associated with guanine nucleotide exchange activation for the Rho/Rac family of small GTP binding proteins, resulting in the conversion of the inactive GTPase to the active form capable of transducing signals. The PH domain has multiple functions. Therefore, these isoforms function as scaffolding proteins to coordinate a Rho signaling pathway and, in addition, function as protein kinase A-anchoring proteins. [provided by RefSeq]
Sequence: MKLNPQQAPLYGDCVVTVLLAEEDKAEDDVVFYLVFLGSTLRHCTSTRKVSSDTLETIAPGHDCCETVKVQLCASKEGLPVFVVAEEDFHFVQDEAYDAAQFLATSAGNQSpecifications
NP_006729 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKLNPQQAPLYGDCVVTVLLAEEDKAEDDVVFYLVFLGSTLRHCTSTRKVSSDTLETIAPGHDCCETVKVQLCASKEGLPVFVVAEEDFHFVQDEAYDAAQFLATSAGNQ | |
RUO | |
AKAP13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11214 | |
AKAP13 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AKAP-Lbc/ARHGEF13/BRX/FLJ11952/FLJ43341/HA-3/Ht31/LBC/PROTO-LB/PROTO-LBC/c-lbc | |
AKAP13 | |
Recombinant | |
wheat germ expression system |