missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AKAP10 Full-length ORF (NP_009133.2, 1 a.a. - 662 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011216-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123926

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein interacts with both the type I and type II regulatory subunits of PKA; therefore, it is a dual-specific AKAP. This protein is highly enriched in mitochondria. It contains RGS (regulator of G protein signalling) domains, in addition to a PKA-RII subunit-binding domain. The mitochondrial localization and the presence of RGS domains may have important implications for the function of this protein in PKA and G protein signal transduction. [provided by RefSeq]

Sequence: MRGAGPSPRQSPRTLRPDPGPAMSFFRRKVKGKEQEKTSDVKSIKASISVHSPQKSTKNHALLEAAGPSHVAINAISANMDSFSSSRTATLKKQPSHMEAAHFGDLGRSCLDYQTQETKSSLSKTLEQVLHDTIVLPYFIQFMELRRMEHLVKFWLEAESFHSTTWSRIRAHSLNTVKQSSLAEPVSPSKKHETTASFLTDSLDKRLEDSGSAQLFMTHSEGIDLNNRTNSTQNHLLLSQECDSAHSLRLEMARAGTHQVSMETQESSSTLTVASRNSPASPLKELSGKLMKSIEQDAVNTFTKYISPDAAKPIPITEAMRNDIIARICGEDGQVDPNCFVLAQSIVFSAMEQEHFSEFLRSHHFCKYQIEVLTSGTVYLADILFCESALFYFSEYMEKEDAVNILQFWLAADNFQSQLAAKKGQYDGQEAQNDAMILYDKYFSLQATHPLGFDDVVRLEIESNICREGGPLPNCFTTPLRQAWTTMEKVFLPGFLSSNLYYKYLNDLIHSVRGDEFLGGNVSLTAPGSVGPPDESHPGSSDSSASQSSVKKASIKILKNFDEAIIVDAASLDPESLYQRTYAGKMTFGRVSDLGQFIRESEPEPDVRKSKGSMFSQAMKKWVQGNTDEAQEELAWKIAKMIVSDIMQQAQYDQPLEKSTKL
Specifications

Specifications

NP_009133.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
100.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
AKAP10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
11216
AKAP10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
D-AKAP2/MGC9414/PRKA10
AKAP10
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers