missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AGTRAP Partial ORF (NP_065083, 108 a.a. - 159 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057085-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138976

  • 3602.46 NOK / 10µg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a transmembrane protein localized to the plasma membrane and perinuclear vesicular structures. The gene product interacts with the angiotensin II type I receptor and negatively regulates angiotensin II signaling. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: HMYRERGGELLVHTGFLGSSQDRSAYQTIDSAEAPADPFAVPEGRSQDARGS
Specifications

Specifications

NP_065083
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.46kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HMYRERGGELLVHTGFLGSSQDRSAYQTIDSAEAPADPFAVPEGRSQDARGS
RUO
AGTRAP
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
57085
AGTRAP (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATRAP/MGC29646
AGTRAP
Yes
Videos
SDS
Documents
Certificates