missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AGPAT7 Full-length ORF (NP_705841.2, 1 a.a. - 524 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00254531-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189643

  • 4115.00 NOK / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Members of the 1-acylglycerol-3-phosphate O-acyltransferase (EC 2.3.1.51) family, such as AGPAT7, catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA), a precursor in the biosynthesis of all glycerolipids. Both LPA and PA are involved in signal transduction (Ye et al., 2005 [PubMed 16243729]).[supplied by OMIM]

Sequence: MSQGSPGDWAPLDPTPGPPASPNPFVHELHLSRLQRVKFCLLGALLAPIRVLLAFIVLFLLWPFAWLQVAGLSEEQLQEPITGWRKTVCHNGVLGLSRLLFFLLGFLRIRVRGQRASRLQAPVLVAAPHSTFFDPIVLLPCDLPKVVSRAENLSVPVIGALLRFNQAILVSRHDPASRRRVVEEVRRRATSGGKWPQVLFFPEGTCSNKKALLKFKPGAFIAGVPVQPVLIRYPNSLDTTSWAWRGPGVLKVLWLTASQPCSIVDVEFLPVYHPSPEESRDPTLYANNVQRVMAQALGIPATECEFVGSLPVIVVGRLKVALEPQLWELGKVLRKAGLSAGYVDAGAEPGRSRMISQEEFARQLQLSDPQTVAGAFGYFQQDTKGLVDFRDVALALAALDGGRSLEELTRLAFELFAEEQAEGPNRLLYKDGFSTILHLLLGSPHPAATALHAELCQAGSSQGLSLCQFQNFSLHDPLYGKLFSTYLRPPHTSRGTSQTPNASSPGNPTALANGTVQAPKQKGD
Specifications

Specifications

NP_705841.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
83.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AGPAT7/AYTL3/FLJ10257/LPAAT-eta/LPEAT2
LPCAT4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
254531
AGPAT7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSQGSPGDWAPLDPTPGPPASPNPFVHELHLSRLQRVKFCLLGALLAPIRVLLAFIVLFLLWPFAWLQVAGLSEEQLQEPITGWRKTVCHNGVLGLSRLLFFLLGFLRIRVRGQRASRLQAPVLVAAPHSTFFDPIVLLPCDLPKVVSRAENLSVPVIGALLRFNQAILVSRHDPASRRRVVEEVRRRATSGGKWPQVLFFPEGTCSNKKALLKFKPGAFIAGVPVQPVLIRYPNSLDTTSWAWRGPGVLKVLWLTASQPCSIVDVEFLPVYHPSPEESRDPTLYANNVQRVMAQALGIPATECEFVGSLPVIVVGRLKVALEPQLWELGKVLRKAGLSAGYVDAGAEPGRSRMISQEEFARQLQLSDPQTVAGAFGYFQQDTKGLVDFRDVALALAALDGGRSLEELTRLAFELFAEEQAEGPNRLLYKDGFSTILHLLLGSPHPAATALHAELCQAGSSQGLSLCQFQNFSLHDPLYGKLFSTYLRPPHTSRGTSQTPNASSPGNPTALANGTVQAPKQKGD
RUO
LPCAT4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers