Learn More
Abnova™ Human AGPAT6 Partial ORF (NP_848934.1, 39 a.a. - 137 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00137964-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Lysophosphatidic acid acyltransferases (EC 2.3.1.51) catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.[supplied by OMIM]
Sequence: SFGIRKLYMKSLLKIFAWATLRMERGAKEKNHQLYKPYTNGIIAKDPTSLEEEIKEIRRSGSSKALDNTPEFELSDIFYFCRKGMETIMDDEVTKRFSASpecifications
NP_848934.1 | |
Liquid | |
137964 | |
AGPAT6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586M1819/LPAAT-zeta/LPAATZ/TSARG7 | |
AGPAT6 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SFGIRKLYMKSLLKIFAWATLRMERGAKEKNHQLYKPYTNGIIAKDPTSLEEEIKEIRRSGSSKALDNTPEFELSDIFYFCRKGMETIMDDEVTKRFSA | |
RUO | |
AGPAT6 | |
Wheat Germ (in vitro) | |
GST |