Learn More
Abnova™ Human AGA Full-length ORF (AAH12392, 1 a.a. - 346 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000175-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Aspartylglucosaminidase is involved in the catabolism of N-linked oligosaccharides of glycoproteins. It cleaves asparagine from N-acetylglucosamines as one of the final steps in the lysosomal breakdown of glycoproteins. The lysosomal storage disease aspartylglycosaminuria is caused by a deficiency in the AGA enzyme. [provided by RefSeq]
Sequence: MARKSNLPVLLVPFLLCQALVRCSSPLPLVVNTWPFKNATEAAWRALASGGSALDAVESGCAMCEREQCDGSVGFGGSPDELGETTLDAMIMDGTTMDVGAVGDLRRIKNAIGVARKVLEHTTHTLLVGESATTFAQSMGFINEDLSTSASQALHSDWLARNCQPNYWRNVIPDPSKYCGPYKPPGILKQDIPIHKETEDDRGHDTIGMVVIHKTGHIAAGTSTNGIKFKIHGRVGDSPIPGAGAYADDTAGAAAATGNGDILMRFLPSYQAVEYMRRGEDPTIACQKVISRIQKHFPEFFGAVICANVTGSYGAACNKLSTFTQFSFMVYNSEKNQPTEEKVDCISpecifications
AAH12392 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
AGA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
175 | |
AGA (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGU/ASRG/GA | |
AGA | |
Recombinant | |
wheat germ expression system |