Learn More
Abnova™ Human AES Full-length ORF (NP_945320.1, 1 a.a. - 264 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000166-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is similar in sequence to the amino terminus of Drosophila enhancer of split groucho, a protein involved in neurogenesis during embryonic development. The encoded protein, which belongs to the groucho/TLE family of proteins, can function as a homooligomer or as a heteroologimer with other family members to dominantly repress the expression of other family member genes. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MCHKNGFPQEGGITAAFLQKRKLRLSKNHRPARAKVTEHVRGTRPGRATAGPAASTRAAGSLFFDRWGNRGPAGCRGSSHLPQQLKFTTSDSCDRIKDEFQLLQAQYHSLKLECDKLASEKSEMQRHYVMYYEMSYGLNIEMHKQAEIVKRLNGICAQVLPYLSQEHQQQVLGAIERAKQVTAPELNSIIRQQLQAHQLSQLQALALPLTPLPVGLQPPSLPAVSAGTGLLSLSALGSQAHLSKEDKNGHDGDTHQEDDGEKSDSpecifications
NP_945320.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
AES | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
166 | |
AES (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AES-1/AES-2/ESP1/GRG/GRG5/TLE5 | |
AES | |
Recombinant | |
wheat germ expression system |