Learn More
Abnova™ Human ADSL Full-length ORF (NP_000017.1, 1 a.a. - 484 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000158-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Adenylsuccinate lyase is involved in both de novo synthesis of purines and formation of adenosine monophosphate from inosine monophosphate. It catalyzes two reactions in AMP biosynthesis: the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and removal of fumarate from adenylosuccinate to give AMP. Adenylosuccinase deficiency results in succinylpurinemic autism, psychomotor retardation, and , in some cases, growth retardation associated with muscle wasting and epilepsy. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MAAGGDHGSPDSYRSPLASRYASPEMCFVFSDRYKFRTWRQLWLWLAEAEQTLGLPITDEQIQEMKSNLENIDFKMAAEEEKRLRHDVMAHVHTFGHCCPKAAGIIHLGATSCYVGDNTDLIILRNALDLLLPKLARVISRLADFAKERASLPTLGFTHFQPAQLTTVGKRCCLWIQDLCMDLQNLKRVRDDLRFRGVKGTTGTQASFLQLFEGDDHKVEQLDKMVTEKAGFKRAFIITGQTYTRKVDIEVLSVLASLGASVHKICTDIRLLANLKEMEEPFEKQQIGSSAMPYKRNPMRSERCCSLARHLMTLVMDPLQTASVQWFERTLDDSANRRICLAEAFLTADTILNTLQNISEGLVVYPKVIERRIRQELPFMATENIIMAMVKAGGSRQDCHEKIRVLSQQAASVVKQEGGDNDLIERIQVDAYFSPIHSQLDHLLDPSSFTGRASQQVQRFLEEEVYPLLKPYESVMKVKAELCLSpecifications
NP_000017.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ADSL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
158 | |
ADSL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMPS/ASASE/ASL | |
ADSL | |
Recombinant | |
wheat germ expression system |