Learn More
Abnova™ Human ADCY2 Partial ORF (NP_065433, 977 a.a. - 1086 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000108-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex. [provided by RefSeq]
Sequence: GKLDAINKHSFNDFKLRVGINHGPVIAGVIGAQKPQYDIWGNTVNVASRMDSTGVLDKIQVTEETSLVLQTLGYTCTCRGIINVKGKGDLKTYFVNTEMSRSLSQSNVASSpecifications
NP_065433 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GKLDAINKHSFNDFKLRVGINHGPVIAGVIGAQKPQYDIWGNTVNVASRMDSTGVLDKIQVTEETSLVLQTLGYTCTCRGIINVKGKGDLKTYFVNTEMSRSLSQSNVAS | |
RUO | |
ADCY2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
108 | |
ADCY2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AC2/FLJ16822/FLJ45092/HBAC2/KIAA1060/MGC133314 | |
ADCY2 | |
Recombinant | |
wheat germ expression system |