Learn More
Abnova™ Human ADAT3 Full-length ORF (NP_612431.1, 1 a.a. - 351 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand:  Abnova™ H00113179-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16126983
  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
Description
Sequence: MEPAPGLVEQPKCLEAGSPEPEPAPWQALPVLSEKQSGDVELVLAYAAPVLDKRQTSRLLKEVSALHPLPAQPHLKRVRPSRDAGSPHALEMLLCLAGPASGPRSLAELLPRPAVDPRGLGQPFLVPVPARPPLTRGQFEEARAHWPTSFHEDKQVTSALAGRLFSTQERAAMQSHMERAVWAARRAAARGLRAVGAVVVDPASDRVLATGHDCSCADNPLLHAVMVCVDLVARGQGRGTYDFRPFPACSFAPAAAPQAVRAGAVRKLDADEDGLPYLCTGYDLYVTREPCAMCAMALVHARILRVFYGAPSPDGALGTRFRIHARPDLNHRFQVFRGVLEEQCRWLDPDT
Specifications
Specifications
NP_612431.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ADAT3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
113179
ADAT3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FWP005/MST121/MSTP121/S863-5/TAD3
ADAT3
Recombinant
wheat germ expression system