Learn More
Abnova™ Human ADAMTS2 Partial ORF (NP_055059, 1112 a.a. - 1210 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009509-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene excises the N-propeptide of type I, type II and type V procollagens. Mutations in this gene cause Ehlers-Danlos syndrome type VIIC, a recessively inherited connective-tissue disorder. Alternative splicing results in two transcript variants. The short transcript encodes a protein which has no significant procollagen N-peptidase activity. [provided by RefSeq]
Sequence: KHNDIDVFMPTLPVPTVAMEVRPSPSTPLEVPLNASSTNATEDHPETNAVDEPYKIHGLEDEVQPPNLIPRRPSPYEKTRNQRIQELIDEMRKKEMLGKSpecifications
NP_055059 | |
Liquid | |
9509 | |
ADAMTS2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADAM-TS2/ADAMTS-3/NPI/PCINP/PCPNI/hPCPNI | |
ADAMTS2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KHNDIDVFMPTLPVPTVAMEVRPSPSTPLEVPLNASSTNATEDHPETNAVDEPYKIHGLEDEVQPPNLIPRRPSPYEKTRNQRIQELIDEMRKKEMLGK | |
RUO | |
ADAMTS2 | |
Wheat Germ (in vitro) | |
GST |