Learn More
Abnova™ Human ACTL7B Full-length ORF (NP_006677.1, 1 a.a. - 415 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010880-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of a family of actin-related proteins (ARPs) which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene (ACTL7B), and related gene, ACTL7A, are intronless, and are located approximately 4 kb apart in a head-to-head orientation within the familial dysautonomia candidate region on 9q31. Based on mutational analysis of the ACTL7B gene in patients with this disorder, it was concluded that it is unlikely to be involved in the pathogenesis of dysautonomia. Unlike ACTL7A, the ACTL7B gene is expressed predominantly in the testis, however, its exact function is not known. [provided by RefSeq]
Sequence: MATRNSPMPLGTAQGDPGEAGTRPGPDASLRDTGAATQLKMKPRKVHKIKAVIIDLGSQYCKCGYAGEPRPTYFISSTVGKRCPEAADAGDTRKWTLVGHELLNTEAPLKLVNPLKHGIVVDWDCVQDIWEYIFRTAMKILPEEHAVLVSDPPLSPSSNREKYAELMFETFGIPAMHVTSQSLLSIYSYGKTSGLVVESGHGVSHVVPISEGDVLPGLTSRADYAGGDLTNYLMQLLNEAGHAFTDDHLHIIEHIKKKCCYAAFLPEEELGLVPEELRVDYELPDGKLITIGQERFRCSEMLFQPSLAGSTQPGLPELTAACLGRCQDTGFKEEMAANVLLCGGCTMLDGFPERFQRELSLLCPGDSPAVAAAPERKTSVWTGGSILASLQAFQQLWVSKEEFEERGSVAIYSKCSpecifications
NP_006677.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ACTL7B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10880 | |
ACTL7B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACTL7B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |