Learn More
Abnova™ Human ABCG1 Partial ORF (AAH29158, 21 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009619-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the White subfamily. It is involved in macrophage cholesterol and phospholipids transport, and may regulate cellular lipid homeostasis in other cell types. Six alternative splice variants have been identified. [provided by RefSeq]
Sequence: AEMTEPKSVCVSVDEVVSSNMEATETDLLNGHLKKVDNNLTEAQRFSSLPRRAAVNIEFRDLSYSVPEGPWWRKKGYKTLLKGISGKFNSGELVAIMGPSGAGKSTLMNISpecifications
AAH29158 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AEMTEPKSVCVSVDEVVSSNMEATETDLLNGHLKKVDNNLTEAQRFSSLPRRAAVNIEFRDLSYSVPEGPWWRKKGYKTLLKGISGKFNSGELVAIMGPSGAGKSTLMNI | |
RUO | |
ABCG1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9619 | |
ABCG1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABC8/MGC34313/WHITE1 | |
ABCG1 | |
Recombinant | |
wheat germ expression system |