Learn More
Abnova™ Human ABCD3 Partial ORF (NP_002849.1, 351 a.a. - 449 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005825-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. This peroxisomal membrane protein likely plays an important role in peroxisome biogenesis. Mutations have been associated with some forms of Zellweger syndrome, a heterogeneous group of peroxisome assembly disorders. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: SELLEDYYQSGRMLLRMSQALGRIVLAGREMTRLAGFTARITELMQVLKDLNHGKYERTMVSQQEKGIEGVQVIPLIPGAGEIIIADNIIKFDHVPLATSpecifications
NP_002849.1 | |
Liquid | |
5825 | |
ABCD3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABC43/PMP70/PXMP1 | |
ABCD3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SELLEDYYQSGRMLLRMSQALGRIVLAGREMTRLAGFTARITELMQVLKDLNHGKYERTMVSQQEKGIEGVQVIPLIPGAGEIIIADNIIKFDHVPLAT | |
RUO | |
ABCD3 | |
Wheat Germ (in vitro) | |
GST |