Learn More
HUBER HTS PS1 Heat Exchanger System
Product Code. 15226507
Click to view available options
Flow Rate:
8 L/min.
Unit Size:
Each
Product Code. 15226507
Brand: HUBER 3068.0001.00
Only null left
Specifications
Ambient Temperature 5°C to 40°C
Flow Rate 8 L/min.
Pump Type Circulation
Temperature Range 5°C to 80°C
Max. Pressure 0.2 bar
Model HTS 1
Width (Metric) 280 mm
Height (Metric) 387 mm
Depth (Metric) 398 mm
Weight (Metric) 18 kg
Electrical Requirements 200-240 V 1∼/2 ∼ 50/60 Hz
Voltage 220/240 V
Frequency 50/60 Hz