missing translation for 'onlineSavingsMsg'
Learn More

HTATSF1 Polyclonal Antibody, Invitrogen™

Product Code. 17187763
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17187763 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17187763 Supplier Invitrogen Supplier No. PA5109836

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MSGTNLDGND EFDEQLRMQE LYGDGKDGDT QTDAGGEPDS LGQQPTDTPY EWDLDKKAWF PKITEDFIAT YQANYGFSND GASSSTANVE DVHARTAEEP PQEKAPEPTD ARKKGEKRKA ESGWFHVEED RNTNVYVSGL PPDITVDEFI QLMSKFGIIM RDPQTEEFKV KLYKDNQGNL

The protein encoded by this gene functions as a cofactor for the stimulation of transcriptional elongation by HIV-1 Tat, which binds to the HIV-1 promoter through Tat-TAR interaction. This protein may also serve as a dual-function factor to couple transcription and splicing and to facilitate their reciprocal activation. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTATSF1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Htatsf1
Gene Accession No. O43719, Q8BGC0
Gene Alias 1600023H17Rik; 2600017A12Rik; 2700077B20Rik; cofactor required for Tat activation of HIV-1 transcription; dJ196E23.2; HIV TAT specific factor 1; HIV Tat-specific factor 1; HIV Tat-specific factor 1 homolog; HIV-1 Tat specific factor 1; Htatsf1; RGD1563377; RP1-196E23.2; TATSF1; Tat-SF1
Gene Symbols Htatsf1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human HTATSF1 (NP_055315.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27336, 317612, 72459
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.