Learn More
Atago™ HSR–500 Hand-Held Refractometer
Description
- Concentation via refractive index measurement
Specifications
Specifications
| Type | Handheld Refractometer |
| Scale Range | 0 to 90 Brix |
| Model | HSR–500 |
| Scale Type | Brix |
| Weight (Metric) | 600 g |
| Dimensions (L x W x H) | 4 x 4 x 20 cm |