Description
Western blot analysis using this antibody detected a band around 110 kDa in various lysates.
HSPH1 prevents the aggregation of denatured proteins in cells under severe stress, on which the ATP levels decrease markedly. This protein inhibits HSPA8/HSC70 ATPase and chaperone activities.
Specifications
Specifications
| Antigen | HSPH1 |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3H5A5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | HSPH1 |
| Gene Accession No. | Q92598 |
| Gene Alias | Antigen NY CO 25, DKFZp686M05240, Heat shock 110 kDa protein, Heat shock protein 105 kDa, HSP105, HSP105A, HSP105B, HSP110, HSPH1, KIAA0201, NY CO 25 |
| Show More |