missing translation for 'onlineSavingsMsg'
Learn More

HSPB11 Mouse anti-Human, Clone: 2D11B3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68059-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17619726

  • 4593.76 NOK / 150µL
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HSPB11 Monoclonal antibody apecifically detetects HSPB11 antigen in Human Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

HSPB11
Monoclonal
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
HSPB11
HSPB11 fusion protein Ag8804
150 μL
51668
Store at -20°C. Stable for one year after shipment.
IgG2a
Western Blot, ELISA
2D11B3
Unconjugated
BC005245
Mouse
Protein A purification
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers