missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA1L Mouse anti-Human, Mouse, Rat, Clone: 3E7H2, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Especificaciones
| Antigen | HSPA1L |
|---|---|
| Clone | 3E7H2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|
16808975
|
Proteintech
66780-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898965
|
Proteintech
66780-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
This antibody well recognizes the endogenous HSPA1L protein in multiple cell lines.
In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock proteins. One class of stress proteins, termed the "small heat shock proteins" is comprised of a diverse group of proteins from ∽15 to >30 kDa, with a single form found in yeast and human cells and multiple related forms found in higher plants. A direct relationship between elevated stress protein expression and neoplasia has been suggested from studies on estrogen-induced proteins in breast cancer cells. In addition, Hsp27 exhibits rapid increased phosphorylation in response to various mitogens, tumor promoter (e. g. phorbol esters) and calcium ionophores. The expression of a 24 kDa protein was shown to correlate with steroid hormone receptor in cell lines, organs, and tumors. The 24 kDa estrogen-induced protein was found to be expressed at high levels in carcinoma of the breast. Another study has suggested that levels of the Hsp27 correlate with endometrial adenocarcinomas. Interestingly, the regulation of human Hsp27 by estrogens and heat shock is reminiscent of the ecdysteroid and heat shock activation of the drosophila Hsp20 genes.Especificaciones
| HSPA1L | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Heat shock 70 kDa protein 1L, HSP70 1L, HSP70 HOM, HSP70T, HSPA1L, hum70t | |
| Hspa1l | |
| IgG2b | |
| Protein A | |
| Antibody |
| 3E7H2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Human, Rat | |
| P16627, P34931, P55063 | |
| 15482, 24963, 3305 | |
| HSPA1L Fusion Protein Ag5182 | |
| Primary | |
| -20°C | |
| HSPA1L |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto