missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HSP90 alpha Antibody (HSP90AA1/7426) - Azide and BSA Free, Novus Biologicals™

Product Code. 30030366 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
30030366 100 μg 100µg
1 options
This item is not returnable. View return policy

Product Code. 30030366

Brand: Novus Biologicals™ NBP326952

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HSP90 alpha Monoclonal antibody specifically detects HSP90 alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP90 alpha
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HSP90AA1/7426
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Formulation 10 mM PBS
Gene Alias FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Host Species Mouse
Immunogen Recombinant fragment (around aa500-700) of human HSP90 alpha protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3320
Target Species Human
Content And Storage Store at -20°C to -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.