Learn More
HSP47 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10875-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16847553
  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
Description
SERPINH1 is a member of the serpin superfamily of serine proteinase inhibitors, and its expression is induced by heat shock. The SERPINH1 protein localizes to the endoplasmic reticulum lumen and binds collagen; thus it is thought to be a molecular chaperone involved in the maturation of collagen molecules. Autoantibodies to SERPINH1 have been found in patients with rheumatoid arthritis.
Specifications
Specifications
HSP47
Polyclonal
Unconjugated
SERPINH1
47 kDa heat shock protein, AsTP3, CBP1, CBP2, Collagen binding protein, Colligin, gp46, HSP47, PIG14, PPROM, RA A47, Serpin H1, SERPINH1, SERPINH2
Rabbit
Antigen Affinity Chromatography
RUO
12406, 871
-20°C
Liquid
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P19324, P50454
SERPINH1
HSP47 Fusion Protein Ag1301
20 μL
Primary
Human, Mouse