missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSP40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517382
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other HSPs.
TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.
Specifications
| HSP40 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB1 | |
| 0610007I11Rik; DjB1; DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ heat shock protein family (Hsp40) member B1; dnaJ homolog subfamily B member 1; DnaJ protein homolog 1; DNAJ1; Dnajb1; HDJ1; hDj-1; Heat shock 40 kDa protein 1; Heat shock protein; HSP; HSP 40; HSP40; HSPF1; human DnaJ protein 1; NDAJB1; radial spoke 16 homolog B; RSPH16B; Sis1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3337, 361384, 81489 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 35 μg/mL | |
| 0.01M HEPES with 50% glycerol, 100 μg/mL BSA, 0.15M NaCl and no preservative; pH 7.5 | |
| P25685, Q9QYJ3 | |
| DNAJB1 | |
| Synthetic peptide corresponding to Glu300 of human HSP40/Hdj1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction