missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-25150
This item is not returnable.
View return policy
Description
HSP27 Polyclonal specifically detects HSP27 in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| HSP27 | |
| Polyclonal | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000 | |
| P04792 | |
| HSPB1 | |
| Recombinant full length human HSP27 expressed in and purified from E. coli | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Bovine, Porcine, Mouse, Rat and other Mammals. Other species have not been tested. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgY |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Supplied as an aliquot of IgY preparation with 5mM Sodium Azide | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| Chicken | |
| 27 kDa | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
| 3315 | |
| Human, Rat, Mouse (Negative) | |
| Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction