Learn More
HSP20 Mouse anti-Human, Unconjugated, Clone: 1G7D2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67327-1-IG-150UL
Description
The HSP20 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HSP20. This antibody recognizes human, mouse, rat, pig antigen. The HSP20 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| HSP20 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, ELISA | |
| 1G7D2 | |
| Unconjugated | |
| 150 μL | |
| 126393 | |
| IgG2a |