missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSF4 Polyclonal Antibody, Invitrogen™

Product Code. 17960653
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17960653 200 μL 200µL
17919712 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17960653 Supplier Invitrogen™ Supplier No. PA5145407

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSF4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene HSF4
Gene Accession No. Q9R0L1, Q9ULV5
Gene Alias ATHSF4; AT-HSFB1; binds heat shock DNA elements; cataract; CTM; CTRCT5; DNA binding protein; heat shock factor 4; heat shock factor protein 4; Heat shock transcription factor 4; hHSF4; HSF 4; Hsf4; HSFB1; HSTF 4; HSTF4; ldis1; Marner; mHSF4
Gene Symbols HSF4
Host Species Rabbit
Immunogen Recombinant Human HSF4 protein, Val171-Arg284 + SPPATACGCGAGHPGRERELQPRGAQECPTA (Accession #Q9ULV5).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26386, 3299
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.