missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | HSF1 |
|---|---|
| Concentration | 0.52 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16846305
|
Proteintech
51034-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16836305
|
Proteintech
51034-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5′ UTR, but encoding the same protein, have been found for this gene.Specifications
| HSF1 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| P38532, Q00613 | |
| 15499, 3297, 79245 | |
| HSF1 Fusion Protein Ag0739 | |
| Primary | |
| -20°C | |
| HSF1 |
| 0.52 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Heat shock factor protein 1, HSF 1, HSF1, HSTF 1, HSTF1 | |
| HSF1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title