Learn More
HSF1 Mouse anti-Human, Unconjugated, Clone: 1F10A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67189-1-IG-20UL
This item is not returnable.
View return policy
Description
The HSF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HSF1. This antibody recognizes human, mouse, rat antigen. The HSF1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| HSF1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1F10A5 | |
| Unconjugated | |
| 20 μL | |
| 3297 | |
| IgG1 |