Learn More
HSF1 Mouse anti-Human, Unconjugated, Clone: 1F10A5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67189-1-IG-20UL
 View more versions of this product
Product Code. 16809695
  • 1710.33 NOK / 20µL
Description
The HSF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HSF1. This antibody recognizes human, mouse, rat antigen. The HSF1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
HSF1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1F10A5
Unconjugated
20 μL
3297
IgG1