missing translation for 'onlineSavingsMsg'
Learn More

HSF1 Mouse anti-Human, Unconjugated, Clone: 1F10A5, Proteintech

Product Code. 16899685 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16899685 150 μL 150µL
16809695 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16899685

Brand: Proteintech 671891IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The HSF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HSF1. This antibody recognizes human, mouse, rat antigen. The HSF1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antigen HSF1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Classification Monoclonal
Clone 1F10A5
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 3297
Target Species Human, Rat, Mouse
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.