missing translation for 'onlineSavingsMsg'
Learn More

HSDL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16988-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16846794

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HSDL1 gene ontology annotations related to this gene include oxidoreductase activity.
Specifications

Specifications

HSDL1
Polyclonal
Unconjugated
HSDL1
HSDL1, SDR12C3
Rabbit
Antigen Affinity Chromatography
RUO
361418, 72552, 83693
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3SXM5, Q4V8B7, Q8BTX9
Hsdl1
HSDL1 Fusion Protein Ag10588
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers