Learn More
HSDL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16856794 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16856794 20 μL 20µL
16846794 150 μL 150µL
2 options
Product Code. 16856794
Brand: Proteintech 169881AP20UL
Rabbit Polyclonal Antibody
HSDL1 gene ontology annotations related to this gene include oxidoreductase activity.
Specifications
Antigen HSDL1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HSDL1
Gene Accession No. Q3SXM5, Q4V8B7, Q8BTX9
Gene Alias HSDL1, SDR12C3
Gene Symbols Hsdl1
Host Species Rabbit
Immunogen HSDL1 Fusion Protein Ag10588
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361418, 72552, 83693
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG