missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11855-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene has both oxidoreductase and epimerase activities and is involved in androgen catabolism. The oxidoreductase activity can convert 3 alpha-adiol to dihydrotestosterone, while the epimerase activity can convert androsterone to epi-androsterone. Both reactions use NAD+ as the preferred cofactor. This gene is a member of the retinol dehydrogenase family. Transcript variants utilizing alternative polyadenylation signals exist.Specifications
| HSD17B6 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B6 | |
| 17 beta HSD 6, 17 beta HSD6, beta HSE, HSD17B6, HSE, RODH, SDR9C6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27400, 286964, 8630 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14756, O54753, Q9R092 | |
| Hsd17b6 | |
| HSD17B6 Fusion Protein Ag2427 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction