missing translation for 'onlineSavingsMsg'
Learn More

HSD17B2 Antibody [Alexa Fluor« 750], Novus Biologicals Biologicals™

Product Code. 30499632 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499632 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30499632 Supplier Novus Biologicals Supplier No. NBP335907AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HSD17B2 Polyclonal antibody specifically detects HSD17B2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias 17-beta-hydroxysteroid dehydrogenase type 2, 20 alpha-hydroxysteroid dehydrogenase, E2DH, EC 1.1.1.62,17-beta-HSD 2, EC 1.1.1.63, EDH17B220-alpha-HSD, estradiol 17-beta-dehydrogenase 2, HSD17, hydroxysteroid (17-beta) dehydrogenase 2, Microsomal 17-beta-hydroxysteroid dehydrogenase, SDR9C2, short chain dehydrogenase/reductase family 9C, member 2, Testosterone 17-beta-dehydrogenase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-200 of human HSD17B2 (NP_002144.1).,, Sequence:, GPGAEELRRTCSPRLSVLQMDITKPVQIKDAYSKVAAMLQDRGLWAVINNAGVLGFPTDGELLLMTDYKQCMAVNFFGTVEVTKTFLPLLRKSKGRLVNVSSMGGGAPMERLASYGSSKAAVTMFSSVMRLELSKWGIKVASIQPGGFLTNIAGTSDKWEKLEKDILDHLPAEVQEDYGQDYILAQRNFLLLINSLASKDFSPVLRDIQHAILAKSPFAYYTPGKGAYLWICLAHYLPIGIYDYFAKRHFGQDKPMPRALRMPNYKKKAT
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3294
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.