missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27830-1-AP-150UL
This item is not returnable.
View return policy
Description
HSD17B1 is favors the reduction of estrogens and androgens. It also has 20-alpha-HSD activity. It uses preferentially NADH.Specifications
| HSD17B1 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B1 | |
| 17 beta HSD 1, 20 alpha HSD, E17KSR, E2DH, EDH17B1, EDH17B2, EDHB17, HSD17, HSD17B1, SDR28C1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3292 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P14061 | |
| HSD17B1 | |
| HSD17B1 Fusion Protein Ag27213 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction