missing translation for 'onlineSavingsMsg'
Learn More

HSD17B1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27830-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16845855

  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HSD17B1 is favors the reduction of estrogens and androgens. It also has 20-alpha-HSD activity. It uses preferentially NADH.
Specifications

Specifications

HSD17B1
Polyclonal
Unconjugated
HSD17B1
17 beta HSD 1, 20 alpha HSD, E17KSR, E2DH, EDH17B1, EDH17B2, EDHB17, HSD17, HSD17B1, SDR28C1
Rabbit
Antigen Affinity Chromatography
RUO
3292
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14061
HSD17B1
HSD17B1 Fusion Protein Ag27213
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers