Learn More
HSD11B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10928-1-AP-20UL
This item is not returnable.
View return policy
Description
HSD11B1 is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, HSD11B1 can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children.The protein encoded by this gene is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, the encoded protein can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. Two transcript variants encoding the same protein have been found for this gene.Specifications
| HSD11B1 | |
| Polyclonal | |
| Unconjugated | |
| HSD11B1 | |
| 11 beta HSD1, 11 DH, HDL, HSD11, HSD11B, HSD11B1, HSD11L, SDR26C1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15483, 25116, 3290 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P16232, P28845, P50172 | |
| HSD11B1 | |
| HSD11B1 Fusion Protein Ag1361 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |