Learn More
CellSensor™ HRE-bla HCT-116 Cell Line
CellSensor™ HRE-bla HCT-116 Cell Line contains a beta - lactamase reporter gene under control of the HRE.
Brand:  CellSensor™ K1643
Additional Details : Weight : 0.01000kg
Product Code. 10514234
  • 90600.00 NOK / Each
Estimated Shipment: 06-06-2024
Description
  • Adaptation to hypoxia is critical for tumor survival and growth, and is mediated largely by transcriptional activation of genes that facilitate short - (e.g., glucose transport) and long - term (e.g., angiogenesis) adaptive mechanisms.
  • This coordinated homeostatic response is mediated in large part through the activation of the heterodimeric transcription factor hypoxia - inducible factor 1 (HIF - 1).
  • As oxygen becomes rate limiting, hydroxylation diminishes and HIF - 1α accumulates and heterodimerizes with the constitutively present α - subunit.
  • The binding of this complex to the cognate hypoxia - response element (HRE) results in transcriptional activation of genes containing such elements within promoter or enhancer elements.
  • This cell line is validated for EC50 and Z'; - factor under optimized conditions using deferoxamine (DFO) and Cobalt Chloride.
  • This cell line has also been tested under various experimental conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time.
  • CellSensor ™ HRE-bla HCT - 116 cells were stimulated with deferoxamine (DFO) over the indicated concentration range in a 384 - well format.
  • Cells were incubated for 24 hrs with agonist and 0.5% DMSO and then combined with LiveBLAzer™ - FRET B/G Substrate (CCF4 - AM) for 2 hrs.
  • Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader.
  • The Response Ratios were plotted against the indicated concentrations of DFO.
  • Response ratios were plotted against the indicated concentrations of CoCl2.
Specifications
Specifications
Store in liquid nitrogen immediately upon receipt, or thaw for immediate use
Signaling Pathway
1 Vial
Dividing Cells