Learn More
HPRT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16864044 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16864044 20 μL 20µL
16854044 150 μL 150µL
2 options
Product Code. 16864044
Brand: Proteintech 150591AP20UL
Rabbit Polyclonal Antibody
The protein is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway.
Specifications
Antigen HPRT1
Applications Immunocytochemistry, Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HPRT1
Gene Accession No. P00492, P00493, P27605
Gene Alias HGPRT, HGPRTase, HPRT, HPRT1
Gene Symbols HPRT1
Host Species Rabbit
Immunogen HPRT1 Fusion Protein Ag7044
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15452, 24465, 3251
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG