Learn More
HPRT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15059-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway.Specifications
| HPRT1 | |
| Polyclonal | |
| Unconjugated | |
| HPRT1 | |
| HGPRT, HGPRTase, HPRT, HPRT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15452, 24465, 3251 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00492, P00493, P27605 | |
| HPRT1 | |
| HPRT1 Fusion Protein Ag7044 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |