missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HPR Polyclonal Antibody, Invitrogen™

Product Code. 17713304
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17713304 20 μL 20µL
17703304 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17713304 Supplier Invitrogen™ Supplier No. PA5121903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a haptoglobin-related protein that binds hemoglobin as efficiently as haptoglobin. Unlike haptoglobin, plasma concentration of this protein is unaffected in patients with sickle cell anemia and extensive intravascular hemolysis, suggesting a difference in binding between haptoglobin-hemoglobin and haptoglobin-related protein-hemoglobin complexes to CD163, the hemoglobin scavenger receptor. This protein may also be a clinically important predictor of recurrence of breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene HPR
Gene Accession No. P00739
Gene Alias A-259H10.2; Haptoglobin-related locus; haptoglobin-related protein; HP; HPR
Gene Symbols HPR
Host Species Rabbit
Immunogen Recombinant Haptoglobin Related Protein (HPR), Ser30-Pro85 (Accession P00739).
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3250
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.