missing translation for 'onlineSavingsMsg'
Learn More

HPPH, MedChemExpress

Product Code. 16431977
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431977 50 mg 50mg
16488487 5 mg 5mg
16416296 100 mg 100mg
16413386 25 mg 25mg
16405828 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16431977 Supplier Medchem Express Supplier No. HY13722

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HPPH (Photochlor) is a second generation photosensitizer, which acts as a photodynamic therapy (PDT) agent.

HPPH (Photochlor) is a second generation photosensitizer, which acts as a photodynamic therapy (PDT) agent.

Specifications

Chemical Name or Material HPPH
Physical Form Solid
CAS 149402-51-7
Color Black
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C39H48N4O4
For Use With (Application) Cancer-programmed cell death
Synonym Photochlor
Solubility Information DMSO : 125 mg/mL (196.29 mM; Need ultrasonic)
SMILES OC(CC[C@@H]([C@@H]1C)/C2=C(C3)/C4=C(C(C)=C(/C=C5C(CC)=C(C)C(/C=C6N/C(C(C)=C\6C(C)OCCCCCC)=C\C1=N2)=N/5)N4)C3=O)=O
Molecular Weight (g/mol) 636.82
Formula Weight 636.82
Percent Purity 98.32%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.