missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPd1 Antibody (DHIC2-4A10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-18953SS
This item is not returnable.
View return policy
Description
HPd1 Monoclonal specifically detects HPd1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| HPd1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100 | |
| DHIC2-4A10, HPd1, human pancreatic duct cell | |
| Human pancreatic enriched duct cells | |
| 0.025 mL | |
| Adaptive Immunity, Apoptosis, Cell Biology, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers | |
| Specific for duct cell types. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| DHIC2-4A10 | |
| Unconjugated | |
| Tissue culture supernatant with 0.09% Sodium Azide | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction