Learn More
Epredia™ HP35 Disposable Microtome Blades
Product Code. 11912355 Shop All Epredia Products
Change view
Click to view available options
Coating:
Coated
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Coating Unit Size
11912355 Coated Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11912355 Supplier Epredia™ Supplier No. 3150734
  • ONLINE EXCLUSIVE PRICE
    1360.42 NOK
    985.96 NOK / Pack of 50
    Save 374.46 NOK
    27% Off
 Restricted Item
In Stock
Epredia™ HP35 microtome blades are coated for reliable and thin sectioning.
  • High-profile
  • Increased durability
TRUSTED_SUSTAINABILITY
Specifications
Product Type High Profile Blade
Profile High
For Use With (Application) Designed for high-quality thin sectioning
Disposable Single-use
Coating Coated
Cutting Angle 34°
Thickness (English) 0.01 in.
Thickness (Metric) 0.31 mm