Learn More
HP1 alpha Antibody (RAB-C133) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-20127JF585
Product Code. 30171218
  • 4205.00 NOK / 0.10mL
Description
HP1 alpha Monoclonal antibody specifically detects HP1 alpha in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Mass Spectrometry,ChIP assay (ChIP),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HP1 alpha
Monoclonal
Janelia Fluor 585
Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha
This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX5 protein under non-denaturing conditions.
0.1 mL
Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing
23468
Store at 4°C in the dark.
IgG κ
Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Mass Spectrometry, ChIP Assay, Immunocytochemistry/Immunofluorescence
RAB-C133
50mM Sodium Borate
Rabbit
Protein A purified