missing translation for 'onlineSavingsMsg'
Learn More

HP Mouse anti-Human, Porcine, Clone: 3B4B9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66229-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847915

  • 1415.13 NOK / 20µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Haptoglobin is an a2-globulin plasma protein synthesized in the liver. It is an acute phase protein which aslo acts as a scavenger of hemoglobin from hemolyzed red cells. The haptoglobin monomer consists of two a and two beta chains; the a chanins bay be hp1Fa, hp1Sa, or hp2a types while the beta chain is constant. Three major phenotypes are recognized: HP1-1, Hp2-1 and Hp-2-2, in which the hp1a chain bay be either the F or S type.
Specifications

Specifications

HP
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P00738, Q8SPS7
HP
HP Fusion Protein Ag9927
20 μL
Primary
Porcine, Human
Antibody
IgG2b
Western Blot
3B4B9
Unconjugated
HP
BP, HAPT, haptoglobin, HP, HP2 ALPHA 2, HPA1S
Mouse
Protein A
RUO
3240, 397061
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers