missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15448-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the regulation of cartilage differentiation. It could also be involved in chondrodysplasias or other cartilage disorders.Specifications
HOXC8 | |
Polyclonal | |
Unconjugated | |
HOXC8 | |
homeobox C8, Homeobox protein Hox 3A, Homeobox protein Hox C8, HOX3, HOX3A, HOXC8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15426, 24460, 3224 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09025, P18866, P31273 | |
HOXC8 | |
HOXC8 Fusion Protein Ag7675 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |