missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HOXB13 (Human) Recombinant Protein
Human HOXB13 partial ORF (AAH07092.1, 61 a.a. - 216 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00010481-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: KQCHPCPGVPQGTSPAPVPYGYFGGGYYSCRVSRSSLKPCAQAATLAAYPAETPTAGEEYPSRPTEFAFYPGYPGTYQPMASYLDVSVVQTLGAPGEPRHDSLLPVDSYQSWALAGGWNSQMCCQGEQNPPGPFWKAAFADSSGQHPPDACAFRRG
Specifications
AAH07092.1 | |
homeobox B13 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOXB13 | |
GST |
10481 | |
Wheat germ expression system | |
25 μg | |
PSGD | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |