Learn More
Howard Leight™ Polar Hood™
Product Code. 12677333
Click to view available options
:
Small/Medium
Product Code. 12677333
Brand: Honeywell Safety Products 1016871
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides protection from cold climates while ensuring optimal ear muff attenuation
  • Ideal for airport ground crews, construction workers, oil/gas/petrochemical workers, frozen food production workers, and others exposed to cold weather
  • Seals around face to prevent exposure to the elements
  • Side panels help eliminate gap between earcup and ear, reducing hazardous noise exposure
  • Adjustable design zips up the front for ventilation
  • Compatible with all Howard Leight ear muffs and fits under most hardhats
  • Bright green trim enhances visibility and safety outdoors
  • Fold-away visor