Learn More
Thermo Scientific™ HotStart™ Storage Reaction Tubes
Product Code. 12358212 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
0.5 mL
Unit Size:
Pack of 96
Product Code. Volume (Metric) unitSize
12358212 0.5 mL Pack of 96
1 options
Product Code. 12358212
Brand: Thermo Scientific™ 600211
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Features:
  • Bead is stable at room temperature
  • Assures synchronous reaction startup
  • PCR product can remain in tube for later analysis
  • Hinged caps provide snug seal
  • Presterilized; RNase- and DNase-free
Recommended for: Complex DNA or cDNA, very low copy number targets, more than 30 thermal cycles, multiple PCR or RT-PCR
TRUSTED_SUSTAINABILITY
Specifications
Description Hot Start 50
Volume (Metric) Working 25 to 50 μL
Volume (Metric) 0.5 mL
Product Type Tube